Search
Search
| Gene Symbol: Nvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026171.1 | NP_080447.1 | 4 - 5 | 333 | 95 | |
| GenBank mRNA | AK004676.1 | - | 4 - 5 | 365 | 95 | |
| AK013982.1 | - | 4 - 5 | 332 | 95 | ||
| AK018049.1 | - | 4 - 5 | 329 | 95 | ||
| AK140509.1 | - | 4 - 5 | 364 | 95 | ||
| BC031847.1 | - | 4 - 5 | 316 | 3964470 | 95 | |
Set Membership: |
|