Search
Search

Scrn1| Gene Symbol: Scrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027268.2 | NP_081544.1 | 4 - 5 | 686 | 76 | |
| GenBank mRNA | AK012765.1 | - | 4 - 5 | 714 | 76 | |
| AK032573.1 | - | 4 - 5 | 713 | 76 | ||
| AK050481.1 | - | 4 - 5 | 704 | 76 | ||
| AK082953.1 | - | 4 - 5 | 686 | 76 | ||
| AK089968.1 | - | 4 - 5 | 676 | 76 | ||
| AK129084.1 | - | 4 - 5 | 673 | 76 | ||
| AK145904.1 | - | 4 - 5 | 688 | 76 | ||
| BC023889.1 | - | 4 - 5 | 673 | 5252046 | 76 | |
| EF410099.1 | - | 637 | 76 | |||
Set Membership: |
|