Search
Search
| Gene Symbol: Evl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163394.1 | NP_001156866.1 | 4 - 5 | 907 | 113 | |
| NM_001163395.1 | NP_001156867.1 | 4 - 5 | 659 | 113 | ||
| NM_001163396.2 | NP_001156868.1 | 5 - 6 | 681 | 113 | ||
| NM_007965.3 | NP_031991.3 | 4 - 5 | 907 | 113 | ||
| GenBank mRNA | AF279662.1 | - | 3 - 4 | 418 | 113 | |
| AK134563.1 | - | 4 - 5 | 659 | 114 | ||
| AK137823.1 | - | 4 - 5 | 496 | 113 | ||
| AK151633.1 | - | 5 - 6 | 682 | 113 | ||
| BC059810.1 | - | 4 - 5 | 907 | 6828523 | 113 | |
| BC096017.1 | - | 4 - 5 | 800 | 113 | ||
| U72519.1 | - | 4 - 5 | 670 | 113 | ||
Set Membership: |
|