Search
Search

Timp1| Gene Symbol: Timp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044384.1 | NP_001037849.1 | 3 - 4 | 375 | 86 | |
| NM_011593.2 | NP_035723.2 | 3 - 4 | 372 | 86 | ||
| GenBank mRNA | BC008107.1 | - | 3 - 4 | 277 | 3158222 | 86 |
| BC034260.1 | - | 3 - 4 | 277 | 3158222 | 86 | |
| BC051260.1 | - | 3 - 4 | 257 | 30056565 | 86 | |
| BU756519.1 | - | 3 - 4 | 86 | |||
| CJ055052.1 | - | 3 - 4 | 287 | 86 | ||
| CJ063120.1 | - | 3 - 4 | 373 | 86 | ||
| M17243.1 | - | 3 - 4 | 238 | 86 | ||
| X04684.1 | - | 3 - 4 | 314 | 86 | ||
Set Membership: |
|