Search
Search
| Gene Symbol: Csn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009972.2 | NP_034102.1 | 4 - 5 | 152 | 109 | |
| GenBank mRNA | AK052803.1 | - | 4 - 5 | 149 | 109 | |
| AK052805.1 | - | 4 - 5 | 149 | 109 | ||
| AK085694.1 | - | 4 - 5 | 149 | 109 | ||
| AK085729.1 | - | 4 - 5 | 149 | 109 | ||
| AK142616.1 | - | 4 - 5 | 149 | 109 | ||
| AK142620.1 | - | 4 - 5 | 149 | 109 | ||
| AK142631.1 | - | 4 - 5 | 149 | 109 | ||
| AK142635.1 | - | 4 - 5 | 149 | 109 | ||
| AK164747.1 | - | 4 - 5 | 149 | 109 | ||
| AK164761.1 | - | 4 - 5 | 149 | 109 | ||
| AK164791.1 | - | 4 - 5 | 149 | 109 | ||
| AK164792.1 | - | 4 - 5 | 146 | 109 | ||
| AK164793.1 | - | 4 - 5 | 146 | 109 | ||
| AK164794.1 | - | 4 - 5 | 149 | 109 | ||
| BC013332.1 | - | 4 - 5 | 149 | 3670912 | 109 | |
| BC020356.1 | - | 4 - 5 | 129 | 109 | ||
| BC021153.1 | - | 4 - 5 | 147 | 3670372 | 109 | |
| BC050270.1 | - | 4 - 5 | 129 | 109 | ||
| BC092098.1 | - | 4 - 5 | 126 | 109 | ||
| X04490.1 | - | 4 - 5 | 148 | 109 | ||
Set Membership: |
|