Search
Search

Setd8| Gene Symbol: Setd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030241.2 | NP_084517.2 | 4 - 5 | 580 | 92 | |
| GenBank mRNA | AK010593.1 | - | 3 - 4 | 579 | 92 | |
| AK030904.1 | - | 4 - 5 | 574 | 92 | ||
| AK088097.1 | - | 3 - 4 | 427 | 92 | ||
| BC108333.1 | - | 4 - 5 | 512 | 4978041 | 92 | |
Set Membership: |
|