Search
Search
| Gene Symbol: Parl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005767.4 | NP_001005767.1 | 8 - 9 | 964 | 76 | |
| GenBank mRNA | AK150980.1 | - | 8 - 9 | 972 | 76 | |
| AK151652.1 | - | 8 - 9 | 963 | 76 | ||
| BC083153.1 | - | 8 - 9 | 964 | 30728006 | 76 | |
Set Membership: |
|