Search
Search

IQCB1| Gene Symbol: IQCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002964.4 | XP_004003013.1 | 3 - 4 | 269 | 73 | |
| XM_012095481.3 | XP_011950871.1 | 3 - 4 | 258 | 73 | ||
| XM_012095486.3 | XP_011950876.1 | 3 - 4 | 269 | 73 | ||
| XM_012095487.3 | XP_011950877.1 | 3 - 4 | 269 | 73 | ||
| XM_012095493.3 | XP_011950883.1 | 3 - 4 | 269 | 73 | ||
Set Membership: |
|