Search
Search

RREB1| Gene Symbol: RREB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958471.1 | XP_027814272.1 | 5 - 6 | 3608 | 61 | |
| XM_027958472.1 | XP_027814273.1 | 4 - 5 | 1592 | 61 | ||
| XM_027958473.1 | XP_027814274.1 | 5 - 6 | 3608 | 61 | ||
| XM_027958474.1 | XP_027814275.1 | 5 - 6 | 3608 | 61 | ||
Set Membership: |
|