Search
Search

SYCE2| Gene Symbol: SYCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969546.1 | XP_027825347.1 | 4 - 5 | 593 | 76 | |
| XM_027969547.1 | XP_027825348.1 | 4 - 5 | 603 | 76 | ||
| XM_027969548.1 | XP_027825349.1 | 4 - 5 | 557 | 76 | ||
Set Membership: |
|