Search
Search

RBM19| Gene Symbol: RBM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956555.1 | XP_027812356.1 | 17 - 18 | 2311 | 55 | |
| XM_027956556.1 | XP_027812357.1 | 16 - 17 | 2272 | 55 | ||
| XM_027956557.1 | XP_027812358.1 | 16 - 17 | 2212 | 55 | ||
Set Membership: |
|