Search
Search

ZFP90| Gene Symbol: ZFP90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257522.2 | XP_008255744.1 | 1 - 2 | 208 | 125 | |
| XM_017342319.1 | XP_017197808.1 | 1 - 2 | 139 | 141 | ||
Set Membership: |
|