Search
Search

EML6| Gene Symbol: EML6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017340774.1 | XP_017196263.1 | 4 - 5 | 713 | 72 | |
| XM_017340775.1 | XP_017196264.1 | 4 - 5 | 715 | 72 | ||
| XM_017340776.1 | XP_017196265.1 | 4 - 5 | 715 | 72 | ||
| XM_017340777.1 | XP_017196266.1 | 4 - 5 | 715 | 72 | ||
| XM_017340778.1 | XP_017196267.1 | 4 - 5 | 716 | 72 | ||
Set Membership: |
|