Search
Search

DHX58| Gene Symbol: DHX58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719391.3 | XP_002719437.1 | 10 - 11 | 2026 | 61 | |
| GenBank mRNA | KF640607.1 | - | 10 - 11 | 1752 | 61 | |
| KF640608.1 | - | 10 - 11 | 1752 | 61 | ||
| KF640609.1 | - | 10 - 11 | 1752 | 61 | ||
Set Membership: |
|