Search
Search

TSPAN18| Gene Symbol: TSPAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709068.3 | XP_002709114.1 | 4 - 5 | 431 | 72 | |
| XM_008270135.2 | XP_008268357.1 | 6 - 7 | 617 | 72 | ||
| XM_008270146.2 | XP_008268368.1 | 7 - 8 | 724 | 72 | ||
| XM_017346150.1 | XP_017201639.1 | 6 - 7 | 596 | 72 | ||
Set Membership: |
|