Search
Search

MYOT| Gene Symbol: MYOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255052.2 | XP_008253274.1 | 9 - 10 | 1649 | 64 | |
| XM_017341076.1 | XP_017196565.1 | 9 - 10 | 1647 | 64 | ||
Set Membership: |
|