Search
Search

LOC4341777| Gene Symbol: LOC4341777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788614.2 | XP_015644100.1 | 10 - 11 | 1226 | 89 | |
| GenBank mRNA | AK073262.1 | - | 10 - 11 | 1213 | 89 | |
Set Membership: |
|