Search
Search

SARAF| Gene Symbol: SARAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145102.2 | XP_015000588.1 | 900 | 117 | ||
| XM_028852526.1 | XP_028708359.1 | 533 | 117 | |||
| GenBank mRNA | JU322457.1 | - | 420 | 117 | ||
| JU475967.1 | - | 422 | 117 | |||
| JV045475.1 | - | 313 | 117 | |||
| JV638187.1 | - | 418 | 117 | |||
Set Membership: |
|