Search
Search

SFXN1| Gene Symbol: SFXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260606.1 | NP_001247535.1 | 531 | 85 | ||
| XM_015141431.2 | XP_014996917.1 | 676 | 85 | |||
| XM_015141432.2 | XP_014996918.1 | 781 | 85 | |||
| XM_015141433.2 | XP_014996919.2 | 734 | 85 | |||
| XM_015141434.2 | XP_014996920.1 | 1084 | 85 | |||
| XM_015141435.2 | XP_014996921.1 | 579 | 85 | |||
| XM_015141436.2 | XP_014996922.1 | 614 | 85 | |||
| XM_028849071.1 | XP_028704904.1 | 709 | 85 | |||
| XM_028849072.1 | XP_028704905.1 | 609 | 85 | |||
| XM_028849073.1 | XP_028704906.1 | 735 | 85 | |||
| XM_028849074.1 | XP_028704907.1 | 735 | 85 | |||
| XM_028849075.1 | XP_028704908.1 | 546 | 85 | |||
| XM_028849076.1 | XP_028704909.1 | 498 | 85 | |||
| GenBank mRNA | JU337162.1 | - | 569 | 85 | ||
| JU477895.1 | - | 532 | 85 | |||
| JV048738.1 | - | 531 | 85 | |||
| JV633839.1 | - | 577 | 85 | |||
Set Membership: |
|