Search
Search

SLC13A4| Gene Symbol: SLC13A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134979.2 | XP_014990465.2 | 1094 | 71 | ||
| XM_015134980.2 | XP_014990466.2 | 1094 | 71 | |||
| GenBank mRNA | JV453304.1 | - | 1047 | 71 | ||
| JV540879.1 | - | 1050 | 71 | |||
Set Membership: |
|