Search
Search

MTMR14| Gene Symbol: MTMR14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028844396.1 | XP_028700229.1 | 645 | 60 | ||
| XM_028844397.1 | XP_028700230.1 | 645 | 60 | |||
| XM_028844399.1 | XP_028700232.1 | 645 | 60 | |||
| GenBank mRNA | JU326733.1 | - | 426 | 60 | ||
| JU330654.1 | - | 426 | 60 | |||
| JU491579.1 | - | 434 | 60 | |||
| JV044034.1 | - | 599 | 60 | |||
| JV044035.1 | - | 599 | 60 | |||
Set Membership: |
|