Search
Search

EPN1| Gene Symbol: EPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089860.4 | XP_001089860.3 | 1921 | 110 | ||
| XM_001090090.4 | XP_001090090.3 | 1846 | 110 | |||
| XM_002801451.3 | XP_002801497.3 | 1941 | 110 | |||
| XM_015125131.2 | XP_014980617.2 | 1866 | 110 | |||
| XM_015125133.2 | XP_014980619.2 | 1375 | 110 | |||
| XM_028839063.1 | XP_028694896.1 | 1796 | 110 | |||
| GenBank mRNA | JU334221.1 | - | 1719 | 110 | ||
| JU475334.1 | - | 1800 | 110 | |||
| JU475335.1 | - | 1780 | 110 | |||
| JV542741.1 | - | 1772 | 110 | |||
| JV564960.1 | - | 1729 | 110 | |||
Set Membership: |
|