Search
Search

CCDC66| Gene Symbol: CCDC66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130647.2 | XP_014986133.1 | 1457 | 89 | ||
| XM_015130648.2 | XP_014986134.1 | 1464 | 89 | |||
| XM_015130649.2 | XP_014986135.1 | 1487 | 89 | |||
| XM_015130650.2 | XP_014986136.1 | 1328 | 89 | |||
| XM_015130651.2 | XP_014986137.1 | 1325 | 89 | |||
| XM_015130655.2 | XP_014986141.1 | 1488 | 89 | |||
| XM_015130656.2 | XP_014986142.1 | 1461 | 89 | |||
| XM_015130657.2 | XP_014986143.1 | 1480 | 89 | |||
| XM_015130659.2 | XP_014986145.1 | 1109 | 89 | |||
| XM_015130660.2 | XP_014986146.1 | 1019 | 89 | |||
| XM_028844274.1 | XP_028700107.1 | 1548 | 89 | |||
| XM_028844275.1 | XP_028700108.1 | 1328 | 89 | |||
| XM_028844277.1 | XP_028700110.1 | 1462 | 89 | |||
| XM_028844278.1 | XP_028700111.1 | 1358 | 89 | |||
| XM_028844279.1 | XP_028700112.1 | 1374 | 89 | |||
| XM_028844280.1 | XP_028700113.1 | 974 | 89 | |||
| XM_028844281.1 | XP_028700114.1 | 1457 | 89 | |||
| XM_028844282.1 | XP_028700115.1 | 1468 | 89 | |||
| GenBank mRNA | JU334100.1 | - | 1300 | 89 | ||
| JU475246.1 | - | 1469 | 89 | |||
| JV046813.1 | - | 1428 | 89 | |||
| JV649011.1 | - | 1460 | 89 | |||
Set Membership: |
|