Search
Search

LONRF3| Gene Symbol: LONRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128078.2 | XP_014983564.1 | 2733 | 67 | ||
| XM_015128079.2 | XP_014983565.1 | 2571 | 67 | |||
| XM_015128080.2 | XP_014983566.1 | 2451 | 67 | |||
| GenBank mRNA | JV661747.1 | - | 1234 | 67 | ||
Set Membership: |
|