Search
Search

TNNI2| Gene Symbol: TNNI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001117040.4 | XP_001117040.1 | 551 | 88 | ||
| XM_015113331.2 | XP_014968817.1 | 576 | 88 | |||
| GenBank mRNA | JU492540.1 | - | 205 | 88 | ||
| JV660164.1 | - | 353 | 88 | |||
Set Membership: |
|