Search
Search

RNF115| Gene Symbol: RNF115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266784.1 | NP_001253713.1 | 742 | 99 | ||
| GenBank mRNA | JU321274.1 | - | 862 | 99 | ||
| JU473040.1 | - | 838 | 99 | |||
| JV046774.1 | - | 742 | 99 | |||
Set Membership: |
|