Search
Search

SLC4A10| Gene Symbol: SLC4A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266840.1 | NP_001253769.1 | 723 | 65 | ||
| XM_015110179.2 | XP_014965665.1 | 754 | 65 | |||
| XM_015110185.2 | XP_014965671.1 | 894 | 65 | |||
| XM_028830509.1 | XP_028686342.1 | 754 | 65 | |||
| XM_028830510.1 | XP_028686343.1 | 754 | 65 | |||
| XM_028830511.1 | XP_028686344.1 | 754 | 65 | |||
| XM_028830512.1 | XP_028686345.1 | 754 | 65 | |||
| XM_028830513.1 | XP_028686346.1 | 1221 | 65 | |||
| XM_028830514.1 | XP_028686347.1 | 1186 | 65 | |||
| XM_028830515.1 | XP_028686348.1 | 1087 | 65 | |||
| XM_028830516.1 | XP_028686349.1 | 992 | 65 | |||
| XM_028830517.1 | XP_028686350.1 | 894 | 65 | |||
| XM_028830518.1 | XP_028686351.1 | 894 | 65 | |||
| XM_028830519.1 | XP_028686352.1 | 1221 | 65 | |||
| XM_028830520.1 | XP_028686353.1 | 1221 | 65 | |||
| XM_028830521.1 | XP_028686354.1 | 894 | 65 | |||
| XM_028830522.1 | XP_028686355.1 | 1085 | 65 | |||
| GenBank mRNA | JU335606.1 | - | 723 | 65 | ||
| JU335607.1 | - | 723 | 65 | |||
Set Membership: |
|