Search
Search

DCC| Gene Symbol: DCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121997.2 | XP_014977483.2 | 4862 | 65 | ||
| XM_028837898.1 | XP_028693731.1 | 4862 | 65 | |||
| XM_028837899.1 | XP_028693732.1 | 4862 | 65 | |||
| XM_028837900.1 | XP_028693733.1 | 4859 | 65 | |||
| XM_028837901.1 | XP_028693734.1 | 4856 | 65 | |||
| XM_028837902.1 | XP_028693735.1 | 4853 | 65 | |||
| XM_028837903.1 | XP_028693736.1 | 4802 | 65 | |||
| GenBank mRNA | JU327133.1 | - | 4104 | 65 | ||
| JU339622.1 | - | 4098 | 65 | |||
Set Membership: |
|