Search
Search

AHCYL2| Gene Symbol: AHCYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001091796.4 | XP_001091796.2 | 884 | 61 | ||
| XM_015134885.2 | XP_014990371.1 | 881 | 61 | |||
| XM_015134886.2 | XP_014990372.1 | 1110 | 61 | |||
| XM_015134887.2 | XP_014990373.2 | 972 | 61 | |||
| XM_015134888.2 | XP_014990374.1 | 778 | 61 | |||
| XM_015134889.2 | XP_014990375.2 | 969 | 61 | |||
| XM_015134890.2 | XP_014990376.1 | 775 | 61 | |||
| GenBank mRNA | JU327555.1 | - | 622 | 61 | ||
| JU474920.1 | - | 828 | 61 | |||
| JV646790.1 | - | 858 | 61 | |||
Set Membership: |
|