Search
Search

SLC38A5| Gene Symbol: SLC38A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841898.1 | XP_028697731.1 | 880 | 67 | ||
| XM_028841899.1 | XP_028697732.1 | 1434 | 67 | |||
| XM_028841900.1 | XP_028697733.1 | 702 | 67 | |||
| GenBank mRNA | JU332341.1 | - | 604 | 67 | ||
| JU496378.1 | - | 540 | 67 | |||
| JV487190.1 | - | 619 | 67 | |||
Set Membership: |
|