Search
Search

GAS8| Gene Symbol: GAS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257569.1 | NP_001244498.1 | 806 | 71 | ||
| XM_015126833.2 | XP_014982319.1 | 1007 | 71 | |||
| XM_015126834.2 | XP_014982320.1 | 952 | 71 | |||
| XM_028840439.1 | XP_028696272.1 | 885 | 71 | |||
| XM_028840440.1 | XP_028696273.1 | 885 | 71 | |||
| XM_028840441.1 | XP_028696274.1 | 851 | 71 | |||
| XM_028840442.1 | XP_028696275.1 | 922 | 71 | |||
| GenBank mRNA | JU333938.1 | - | 806 | 71 | ||
| JV046727.1 | - | 802 | 71 | |||
Set Membership: |
|