Search
Search

GOLGA4| Gene Symbol: GOLGA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131367.2 | XP_014986853.2 | 744 | 87 | ||
| XM_015131368.2 | XP_014986854.2 | 744 | 87 | |||
| XM_015131370.2 | XP_014986856.2 | 717 | 87 | |||
| XM_015131371.2 | XP_014986857.2 | 815 | 87 | |||
| XM_015131373.2 | XP_014986859.2 | 618 | 87 | |||
| GenBank mRNA | JU334066.1 | - | 843 | 87 | ||
| JU334067.1 | - | 843 | 87 | |||
| JU475229.1 | - | 827 | 87 | |||
| JV046795.1 | - | 834 | 87 | |||
| JV046796.1 | - | 834 | 87 | |||
| JV712412.1 | - | 856 | 87 | |||
| JV712413.1 | - | 856 | 87 | |||
Set Membership: |
|