Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM189A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015117864.2 | XP_014973350.2 | 1027 | 109 | ||
GenBank mRNA | JU337166.1 | - | 541 | 109 | ||
JU473647.1 | - | 513 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |