Search
Search

SPECC1L| Gene Symbol: SPECC1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257919.2 | NP_001244848.1 | 3239 | 75 | ||
| XM_015149908.2 | XP_015005394.1 | 3197 | 75 | |||
| XM_015149910.2 | XP_015005396.1 | 3112 | 75 | |||
| XM_028827308.1 | XP_028683141.1 | 3141 | 75 | |||
| XM_028827309.1 | XP_028683142.1 | 3133 | 75 | |||
| XM_028827310.1 | XP_028683143.1 | 3088 | 75 | |||
| XM_028827311.1 | XP_028683144.1 | 3199 | 75 | |||
| XM_028827312.1 | XP_028683145.1 | 432 | 75 | |||
| GenBank mRNA | JU322090.1 | - | 3135 | 75 | ||
| JU322091.1 | - | 3239 | 75 | |||
| JU474921.1 | - | 3278 | 75 | |||
| JU474922.1 | - | 3174 | 75 | |||
| JV046580.1 | - | 3274 | 75 | |||
| JV046581.1 | - | 3170 | 75 | |||
Set Membership: |
|