Search
Search

MED29| Gene Symbol: MED29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266003.1 | NP_001252932.1 | 362 | 120 | ||
| GenBank mRNA | JU335350.1 | - | 362 | 120 | ||
| JU335351.1 | - | 362 | 120 | |||
| JU335352.1 | - | 362 | 120 | |||
| JU335353.1 | - | 362 | 120 | |||
| JU476313.1 | - | 362 | 120 | |||
| JV621524.1 | - | 370 | 120 | |||
Set Membership: |
|