Search
Search

CDHR5| Gene Symbol: CDHR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113243.2 | XP_014968729.2 | 1474 | 139 | ||
| XM_015113244.2 | XP_014968730.2 | 1474 | 139 | |||
| XM_015113245.2 | XP_014968731.2 | 1474 | 139 | |||
| XM_015113247.2 | XP_014968733.2 | 1474 | 139 | |||
| XM_028832973.1 | XP_028688806.1 | 1474 | 139 | |||
Set Membership: |
|