Search
Search

PYROXD1| Gene Symbol: PYROXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098386.4 | XP_001098386.2 | 361 | 62 | ||
| XM_028829313.1 | XP_028685146.1 | 481 | 62 | |||
| GenBank mRNA | JU493727.1 | - | 382 | 62 | ||
| JV661837.1 | - | 284 | 62 | |||
Set Membership: |
|