Search
Search

SLC38A11| Gene Symbol: SLC38A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098784.4 | XP_001098784.1 | 796 | 124 | ||
| XM_015110266.2 | XP_014965752.1 | 968 | 124 | |||
| XM_015110267.2 | XP_014965753.1 | 871 | 124 | |||
| XM_028830782.1 | XP_028686615.1 | 709 | 124 | |||
| XM_028830783.1 | XP_028686616.1 | 432 | 124 | |||
Set Membership: |
|