Search
Search

ZNF415| Gene Symbol: ZNF415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124787.2 | XP_014980273.1 | 2394 | 171 | ||
| XM_015124788.2 | XP_014980274.1 | 1767 | 171 | |||
| XM_028839403.1 | XP_028695236.1 | 2282 | 171 | |||
| XM_028839405.1 | XP_028695238.1 | 3621 | 171 | |||
| XM_028839406.1 | XP_028695239.1 | 1103 | 171 | |||
| XM_028839407.1 | XP_028695240.1 | 888 | 171 | |||
| XM_028839408.1 | XP_028695241.1 | 564 | 171 | |||
| XM_028839409.1 | XP_028695242.1 | 1254 | 171 | |||
| XM_028839410.1 | XP_028695243.1 | 593 | 171 | |||
| XM_028839411.1 | XP_028695244.1 | 590 | 171 | |||
Set Membership: |
|