Search
Search

DCBLD2| Gene Symbol: DCBLD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002802684.3 | XP_002802730.2 | 1803 | 86 | ||
| XM_015129372.2 | XP_014984858.1 | 1816 | 86 | |||
| GenBank mRNA | JV043211.1 | - | 1731 | 86 | ||
| JV642412.1 | - | 1822 | 86 | |||