Search
Search

ANKS3| Gene Symbol: ANKS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125547.2 | XP_014981033.2 | 2370 | 86 | ||
| XM_015125549.2 | XP_014981035.2 | 2171 | 86 | |||
| XM_015125550.2 | XP_014981036.2 | 2550 | 86 | |||
| XM_028840678.1 | XP_028696511.1 | 2437 | 86 | |||
| GenBank mRNA | JU325328.1 | - | 86 | |||
| JV456566.1 | - | 124 | 86 | |||
| JV672675.1 | - | 86 | ||||
Set Membership: |
|