Search
Search

DDX54| Gene Symbol: DDX54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111246.4 | XP_001111246.2 | 2247 | 73 | ||
| XM_015152878.2 | XP_015008364.2 | 2247 | 73 | |||
| GenBank mRNA | JU477164.1 | - | 2213 | 73 | ||
| JV661168.1 | - | 2131 | 73 | |||
| JV723168.1 | - | 2216 | 73 | |||