Search
Search

CALCRL| Gene Symbol: CALCRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110509.2 | XP_014965995.1 | 1779 | 118 | ||
| XM_015110510.2 | XP_014965996.1 | 1828 | 118 | |||
| GenBank mRNA | JU322057.1 | - | 1358 | 118 | ||
| KC855759.1 | - | 942 | 118 | |||
Set Membership: |
|