Search
Search

SIRT4| Gene Symbol: SIRT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152956.2 | XP_015008442.2 | 616 | 91 | ||
| XM_015152957.2 | XP_015008443.2 | 1047 | 91 | |||
| GenBank mRNA | JU323018.1 | - | 537 | 91 | ||