Search
Search

LRCH3| Gene Symbol: LRCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132630.2 | XP_014988116.1 | 1563 | 67 | ||
| XM_015132631.2 | XP_014988117.1 | 1563 | 67 | |||
| XM_015132632.2 | XP_014988118.1 | 1563 | 67 | |||
| XM_015132633.2 | XP_014988119.1 | 1563 | 67 | |||
| XM_015132635.2 | XP_014988121.1 | 1563 | 67 | |||
| XM_015132636.2 | XP_014988122.1 | 1563 | 67 | |||
| XM_015132637.2 | XP_014988123.1 | 1563 | 67 | |||
| XM_028843511.1 | XP_028699344.1 | 1563 | 67 | |||
| XM_028843512.1 | XP_028699345.1 | 1563 | 67 | |||
| XM_028843513.1 | XP_028699346.1 | 1563 | 67 | |||
| GenBank mRNA | JV725436.1 | - | 1522 | 67 | ||
Set Membership: |
|