Search
Search

NMT2| Gene Symbol: NMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146530.2 | XP_015002016.1 | 751 | 106 | ||
| XM_015146531.2 | XP_015002017.1 | 760 | 106 | |||
| GenBank mRNA | JU321850.1 | - | 620 | 106 | ||
| JV044994.1 | - | 627 | 106 | |||
| JV665907.1 | - | 623 | 106 | |||
Set Membership: |
|