Search
Search

CLCN2| Gene Symbol: CLCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131432.2 | XP_014986918.1 | 3623 | 76 | ||
| XM_028843900.1 | XP_028699733.1 | 2422 | 76 | |||
| GenBank mRNA | JU321374.1 | - | 2333 | 76 | ||
| JV046144.1 | - | 2319 | 76 | |||
Set Membership: |
|