Search
Search

FBF1| Gene Symbol: FBF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120402.2 | XP_014975888.2 | 1946 | 65 | ||
| XM_015120404.2 | XP_014975890.2 | 1864 | 65 | |||
| XM_015120406.2 | XP_014975892.2 | 1777 | 65 | |||
| XM_015120407.2 | XP_014975893.2 | 1943 | 65 | |||
| XM_015120408.2 | XP_014975894.2 | 1628 | 65 | |||
| XM_028836811.1 | XP_028692644.1 | 1944 | 65 | |||
| XM_028836813.1 | XP_028692646.1 | 1947 | 65 | |||
| XM_028836814.1 | XP_028692647.1 | 1946 | 65 | |||
| GenBank mRNA | JV664033.1 | - | 1598 | 65 | ||
| JV725673.1 | - | 1599 | 65 | |||
Set Membership: |
|