Search
Search
| Gene Symbol: Necab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133415.1 | NP_596906.1 | 7 - 8 | 681 | 60 | |
| XM_006255703.2 | XP_006255765.1 | 5 - 6 | 1747 | 60 | ||
| GenBank mRNA | AF193757.1 | - | 7 - 8 | 681 | 60 | |
Set Membership: |
|