Search
Search
| Gene Symbol: Tert | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053423.1 | NP_445875.1 | 14 - 15 | 3136 | 54 | |
| XM_008758683.1 | XP_008756905.1 | 14 - 15 | 4383 | 54 | ||
| GenBank mRNA | AY539717.1 | - | 14 - 15 | 3136 | 54 | |
| AY539718.1 | - | 14 - 15 | 3118 | 54 | ||
| AY539719.1 | - | 15 - 16 | 3241 | 54 | ||
| AY539720.1 | - | 14 - 15 | 3209 | 54 | ||
Set Membership: |
|